Merck
  • Analysis of ethyl sulfate in raw wastewater for estimation of alcohol consumption and its correlation with drugs of abuse in the city of Barcelona.
Analysis of ethyl sulfate in raw wastewater for estimation of alcohol consumption and its correlation with drugs of abuse in the city of Barcelona.
Journal of chromatography. A (2014-08-12)
Nicola Mastroianni, Miren Lopez de Alda, Damia Barcelo
ABSTRACT
The increasing, generalized consumption of alcohol, especially among young people, generates great concern in our society due to its negative consequences on public health and safety. Besides the traditional, official methods employed for estimation of alcohol consumption, the monitoring of ethyl sulfate (EtS), a urinary biomarker of alcohol ingestion, in raw wastewater has been recently proposed as an additional tool to estimate alcohol use at community level through the so-called sewage epidemiology approach. In the presented study, a fast and reliable analytical method based on ion-pair liquid chromatography-tandem mass spectrometry (LC-MS/MS) has been optimized and further applied to the analysis of EtS in seven 24h composite samples collected along one week at the inlet of a large sewage treatment plant (STP) located in the Barcelona area. EtS was measured in the entire set of analysed samples, with concentrations ranging from 5.5 to 33μg/L, which correspond to an absolute alcohol consumption of around 11,000 (Wednesday) to 25,000 (Sunday) kg/day. The average per capita absolute alcohol consumption calculated was 18mL/day/inhabitant. Moreover, the levels of EtS measured throughout the week showed high correlation with those of some recreational illicit drugs and metabolites, namely, cocaethylene (r(2)=0.9391, n=5), benzoylecgonine (r(2)=0.9252, n=7), ecstasy (r(2)=0.8950, n=7), amphetamine (r(2)=0.8707, n=7) and cocaine (r(2)=0.6425, n=7), measured in the same samples. This study confirms that the analysis of EtS in raw wastewater can be a useful tool for the estimation of alcohol consumption in an anonymous, fast and economic way, and indicates that consumption of alcohol and some illicit drugs occurs often together.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%