Merck
  • Simultaneous determination of tazarotene and its active metabolite tazarotenic acid in minipig plasma by LC-MS/MS and its application in pharmacokinetic study after topical administration of tazarotene gel.
Simultaneous determination of tazarotene and its active metabolite tazarotenic acid in minipig plasma by LC-MS/MS and its application in pharmacokinetic study after topical administration of tazarotene gel.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-01)
Ying Tong, Hong Pan, Chenglong Sun, Xiaofei Xin, Li Ding, Pengcheng Ma
ABSTRACT
To study the systemic exposure of tazarotene formulation after topical administration, a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the simultaneous determination of tazarotene and tazarotenic acid in minipig plasma. Similar extraction recoveries for both analytes were obtained after the plasma samples were acidified by glacial acetic acid (5%) and extracted by ethyl ether-cyclohexane (4:1, v/v). Separation of the analytes was achieved within a short time by the addition of 0.1% formic acid to the mobile phase. Gradient elution was used to avoid the matrix effect. The method was linear over the concentration range of 10-600 pg/mL for both analytes. The data of intra- and inter-run precision and accuracy were lower than 5.2%, 7.3% and 7.3% for both analytes. The developed method can be applied to investigate the transdermal pharmacokinetics and the systemic exposure of tazarotene formulation after topical administration.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Cyclohexane, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%