Merck
  • Improved oral bioavailability of capsaicin via liposomal nanoformulation: preparation, in vitro drug release and pharmacokinetics in rats.
Improved oral bioavailability of capsaicin via liposomal nanoformulation: preparation, in vitro drug release and pharmacokinetics in rats.
Archives of pharmacal research (2014-09-19)
Yuan Zhu, Miaomiao Wang, Jiajia Zhang, Wei Peng, Caleb Kesse Firempong, Wenwen Deng, Qilong Wang, Shicheng Wang, Feng Shi, Jiangnan Yu, Ximing Xu, Weiming Zhang
ABSTRACT
This study innovatively prepared an effective capsaicin-loaded liposome, a nanoformulation with fewer irritants, for oral administration. The in vitro and in vivo properties of the liposomal encapsulation were investigated and the potential possibility of oral administration evaluated. The liposomal agent composed of phospholipid, cholesterol, sodium cholate and isopropyl myristate was prepared using film-dispersion method. A level A in vitro-in vivo correlation (IVIVC) was established for the first time, which demonstrated an excellent IVIVC of both formulated and free capsaicin in oral administration. Physicochemical characterizations including mean particle size, zeta (ζ) potential and average encapsulation efficiency of capsaicin-loaded liposome were found to be 52.2 ± 1.3 nm, -41.5 ± 2.71 mv and 81.9 ± 2.43 %, respectively. In vivo, liposomal encapsulation allowed a 3.34-fold increase in relative bioavailability compared to free capsaicin. The gastric mucosa irritation studies indicated that the liposomal system was a safe carrier for oral administration. These results support the fact that capsaicin, an effective drug for the treatment of neuropathic pain, could be encapsulated in liposome for improved oral bioavailability. The excellent IVIVC of capsaicin-loaded liposome could also be a promising tool in liposomal formulation development with an added advantage of reduced animal testing.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Acetonitrile, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%