Merck
  • Size and surface functionalization of iron oxide nanoparticles influence the composition and dynamic nature of their protein corona.
Size and surface functionalization of iron oxide nanoparticles influence the composition and dynamic nature of their protein corona.
ACS applied materials & interfaces (2014-08-22)
Jonathan Ashby, Songqin Pan, Wenwan Zhong
ABSTRACT
Nanoparticles (NPs) adsorb proteins when in the biological matrix, and the resulted protein corona could affect NP-cell interactions. The corona has a dynamic nature with the adsorbed proteins constantly exchanging with the free proteins in the matrix at various rates. The rapidly exchanging proteins compose the soft corona, which responds more dynamically to environment changes than the hard corona established by the ones with slow exchange rates. In the present study, the corona formed on the superparamagnetic iron oxide NPs (SPIONs) in human serum was studied by flow field-flow fractionation and ultracentrifugation, which rapidly differentiated the corona proteins based on their exchange rates. By varying the surface hydrophobicity of the SPIONs with a core size around 10 nm, we found out that, the more hydrophobic surface ligand attracted proteins with higher surface hydrophobicity and formed a more dynamic corona with a larger portion of the involved proteins with fast exchange rates. Increasing the core diameter of the SPIONs but keeping the surface ligand the same could also result in a more dynamic corona. A brief investigation of the effect on the cellular uptake of SPIONs using one selected corona protein, transferrin, was conducted. The result showed that, only the stably bound transferrin could significantly enhance cellular uptake, while transferrin bound in a dynamic nature had negligible impact. Our study has led to a better understanding of the relationship between the particle properties and the dynamic nature of the corona, which can help with design of nanomaterials with higher biocompatibility and higher efficacy in biosystems for biomedical applications.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%