Merck
  • Study on the anti-gout activity of chlorogenic acid: improvement on hyperuricemia and gouty inflammation.
Study on the anti-gout activity of chlorogenic acid: improvement on hyperuricemia and gouty inflammation.
The American journal of Chinese medicine (2014-11-12)
Zhao-Qing Meng, Zhao-Hui Tang, Yun-Xia Yan, Chang-Run Guo, Liang Cao, Gang Ding, Wen-Zhe Huang, Zhen-Zhong Wang, Kelvin D G Wang, Wei Xiao, Zhong-Lin Yang
ABSTRACT
Gout is a metabolic disorder associated with hyperuricemia resulting in the deposition of monosodium urate (MSU) crystals in joints and tissues. Lowering serum uric acid (Sur) levels and anti-inflammation are highly essential in treating gout. Chlorogenic acid (CA), as one of the most abundant polyphenols in the Chinese medicines, has been rarely reported to have an anti-gout effect. The model of potassium oxonate (PO)-induced hyperuricemia in mice and MSU crystal-induced inflammation in rats has been established in this study. The potential beneficial effects and mechanisms of CA on hyperuricemia and gouty arthritis were elucidated. The results demonstrated that CA significantly decreased the Sur level by inhibiting the xanthine oxidase (XOD) activity but not increasing the urinary uric acid (Uur) level. In addition, CA also exhibited the effect of suppressing paw swelling. Further investigation indicated that CA improved the symptoms of inflammation induced by MSU crystals by inhibiting the production of proinflammatory cytokines including interleukin-1β (IL-1β), interleukin-6 (IL-6), and tumor necrosis factor-α (TNF-α). The present study suggests that CA may have a considerable potential for development as an anti-gouty arthritis agent for clinical application.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Allopurinol, European Pharmacopoeia (EP) Reference Standard
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydride, in paraffin