Merck
  • The influence of phospholipid on the physicochemical properties and anti-tumor efficacy of liposomes encapsulating cisplatin in mice bearing C26 colon carcinoma.
The influence of phospholipid on the physicochemical properties and anti-tumor efficacy of liposomes encapsulating cisplatin in mice bearing C26 colon carcinoma.
International journal of pharmaceutics (2014-07-23)
Seyedeh Hoda Alavizadeh, Ali Badiee, Shiva Golmohammadzadeh, Mahmoud Reza Jaafari
ABSTRACT
SPI-077, cisplatin stealth liposome, is the best illustration of poor cisplatin release from liposomes and the subsequent negligible therapeutic activity. For this reason, optimizing drug release kinetics is desirable. In this report, cisplatin was encapsulated in liposomes composed of different phosphatidylcholines with various phase transition temperatures (Tm) (HSPC, DPPC, DMPC, soy phosphatidylcholine (SPC)), cholesterol and mPEG2000-DSPE. In vitro cytotoxicity studies indicated that lowering Tm of lipids increases cisplatin release; the highest cytotoxicity was observed in SPCs. Cisplatin plasma concentration was also sensitive to the transition temperature. The highest platinum concentration observed after treatment with HSPC and DPPC liposomes, whilst the lowest was observed with SPC. HSPC and DPPC containing liposomes showed the highest therapeutic efficacy and survival with DPPC exhibited better efficacy in mouse model of C26. It seems that DPPC with Tm (41.5°C) nearly, or close to body temperature maintains good drug retention in blood circulation. Upon extravasation through permeable tumor microvasculature, it gradually releases its payload in the tumor area better than HSPC, with a greater Tm of 55°C. Our data suggests, the choice of Tm for lipid mixture directed to a considerable extent the rate of cisplatin elimination from plasma and therapeutic effects.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%