Merck
  • 1H-NMR-based profiling of organic components in leachate from animal carcasses disposal site with time.
1H-NMR-based profiling of organic components in leachate from animal carcasses disposal site with time.
Environmental science and pollution research international (2014-05-14)
Yong-Kook Kwon, Hyun-Whee Bae, Sun Kyoung Shin, Tae-Wan Jeon, Jungju Seo, Geum-Sook Hwang
ABSTRACT
Leachate, generated by the decomposition of animal carcasses, presents many environmental, sanitary, and food safety hazards. However, research on the characteristics of leachate is lacking. In this study, we performed biochemical profiling of leachate from two animal species (pig and cattle) in two soil types (sandy loam and sandy soil) using (1)H-NMR-based profiling, followed by multivariate data analysis. The leachate was collected from a well-controlled artificial burial site over a 31-week period. Principal components analysis (PCA) of the NMR data showed similar patterns between species and soil types. Organic components, including organic acids and phenols, predominated, and their levels increased with time. The methylamine level in leachate from pig carcasses 18 weeks following burial was significantly higher than that from cattle carcasses; leachate from cattle carcasses in sandy soil 1 week after burial contained unique components (specifically ethanol, formate, alanine, N-methylation, and taurine), in contrast with those from sandy loam soil. This study suggests that a NMR-based profiling approach is useful to characterize the organic components in leachate from animal carcasses over time.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, 97%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
p-Cresol, 99%
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1-Methylhydantoin, 97%
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ethanol
SAFC
Taurine
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material