Merck
  • Uliginosin B, a natural phloroglucinol derivative, presents a multimediated antinociceptive effect in mice.
Uliginosin B, a natural phloroglucinol derivative, presents a multimediated antinociceptive effect in mice.
The Journal of pharmacy and pharmacology (2014-08-15)
Eveline Dischkaln Stolz, Diego Rafael Hasse, Gilsane Lino von Poser, Stela M K Rates
ABSTRACT
Investigate the involvement of monoaminergic and glutamatergic systems on the antinociceptive and ataxic effects of uliginosin B, which we have already demonstrated to be a promising molecular scaffold to develop new analgesic drugs. Uliginosin B was obtained from hexane extract of aerial parts of Hypericum polyanthemum by chromatographic methods. Uliginosin B antinociceptive and motor coordination effects were evaluated in mice by using hot-plate (15 and 90 mg/kg, i.p.) and rotarod (90 mg/kg, i.p.) tests, respectively. The mechanism of action was investigated through pretreatments with prazosin 1 mg/kg intraperitoneal (α1 receptor antagonist), yohimbine 5 mg/kg intraperitoneal (α2 receptor antagonist), pCPA 300 mg/kg intraperitoneal (serotonin synthesis inhibitor) and MK-801 0.25 mg/kg intraperitoneal (N-methyl-D-aspartic acid receptor antagonist). The antinociceptive effect of uliginosin B (15 and 90 mg/kg, i.p.) was reduced significantly by pCPA and MK-801. Prazosin and yohimbine improved the antinociceptive effect of the highest dose (90 mg/kg, i.p.) of uliginosin B only. The ataxic effect of uliginosin B (90 mg/kg, i.p.) was completely prevented by pretreatment with pCPA or MK-801, but it was unaffected by pretreatment with prazosin or yohimbine. These data confirm the contribution of monoaminergic neurotransmission as well as provide the first evidence of glutamatergic neurotransmission contribution to the uliginosin B effects.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
(+)-MK-801 hydrogen maleate, powder
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Yohimbine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard