Merck
  • The herbicide linuron inhibits cholesterol biosynthesis and induces cellular stress responses in brown trout.
The herbicide linuron inhibits cholesterol biosynthesis and induces cellular stress responses in brown trout.
Environmental science & technology (2015-01-31)
Tamsyn M Uren Webster, Mandy H Perry, Eduarda M Santos
ABSTRACT
The herbicide linuron is used worldwide, and has been detected in surface waters as well as in food and drinking water. Toxicological studies have reported that linuron acts as an antiandrogen in vitro and in vivo and disrupts mammalian male reproductive function. However, global mechanisms of linuron toxicity are poorly documented. We used RNA-seq to characterize the hepatic transcriptional response of mature male brown trout exposed for 4 days to 1.7, 15.3, and 225.9 μg/L linuron. We identified a striking decrease in the expression of transcripts encoding the majority of enzymes forming the cholesterol biosynthesis pathway. We also measured a very significant decrease in total hepatic cholesterol in fish exposed to 225.9 μg/L linuron and a negative correlation between total cholesterol and linuron treatment concentration. We hypothesize that inhibition of cholesterol biosynthesis may result from the disruption of androgen signaling by linuron. Additionally, there was increased expression of a number of transcripts involved in cellular stress responses, including cyp1a (up to 560-fold), molecular chaperones, and antioxidant enzymes. We found some evidence of similar patterns of transcriptional change in fish exposed to an environmentally relevant concentration of linuron, and further research should investigate the potential for adverse effects to occur following chronic environmental exposure.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Supelco
Linuron, PESTANAL®, analytical standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
USP
Benzocaine, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzocaine, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%