Merck
The frog inner ear: picture perfect?
Journal of the Association for Research in Otolaryngology : JARO (2015-01-30)
Matthew J Mason, Johannes M Segenhout, Ariadna Cobo-Cuan, Patricia M Quiñones, Pim van Dijk
ABSTRACT
Many recent accounts of the frog peripheral auditory system have reproduced Wever's (1973) schematic cross-section of the ear of a leopard frog. We sought to investigate to what extent this diagram is an accurate and representative depiction of the anuran inner ear, using three-dimensional reconstructions made from serial sections of Rana pipiens, Eleutherodactylus limbatus and Xenopus laevis. In Rana, three discrete contact membranes were found to separate the posterior otic (=endolymphatic) labyrinth from the periotic (=perilymphatic) system: those of the amphibian and basilar recesses and the contact membrane of the saccule. The amphibian 'tegmentum vasculosum' was distinguishable as a thickened epithelial lining within a posterior recess of the superior saccular chamber. These features were also identified in Eleutherodactylus, but in this tiny frog the relative proportions of the semicircular canals and saccule resemble those of ranid tadpoles. There appeared to be a complete fluid pathway between the right and left periotic labyrinths in this species, crossing the cranial cavity. Xenopus lacks a tegmentum vasculosum and a contact membrane of the saccule; the Xenopus ear is further distinguished by a lateral passage separating stapes from periotic cistern and a more direct connection between periotic cistern and basilar recess. The basilar and lagenar recesses are conjoined in this species. Wever's diagram of the inner ear of Rana retains its value for diagrammatic purposes, but it is not anatomically accurate or representative of all frogs. Although Wever identified the contact membrane of the saccule, most recent studies of frog inner ear anatomy have overlooked both this and the amphibian tegmentum vasculosum. These structures deserve further attention.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture