Merck
  • The EF-hand calcium-binding protein tescalcin is a potential oncotarget in colorectal cancer.
The EF-hand calcium-binding protein tescalcin is a potential oncotarget in colorectal cancer.
Oncotarget (2014-05-09)
Yun Hee Kang, Seung Ro Han, Jong-Tae Kim, Seon-Jin Lee, Young Il Yeom, Jeong-Ki Min, Chul-Ho Lee, Jae Wha Kim, Suk Ran Yoon, Do-Young Yoon, Kwan Soo Hong, Geum-Sook Hwang, Hee Cheol Kim, Young-Ha Lee, Hee Gu Lee
ABSTRACT
Tescalcin (TESC) is an EF-hand calcium binding protein that is differentially expressed in several tissues, however it is not reported that the expression and functional roles of TESC in colorectal cancer. Levels of messenger RNA (mRNA) and protein expression of TESC in colorectal cancer tissues were assessed using RT-PCR, real time PCR, immunohistochemistry, and clinicopathologic analyses. Quantitative analysis of TESC levels in serum specimens was performed using sandwich ELISA. Colorectal cancer cells transfected with TESC small interfering RNA and short hairpin RNA were examined in cell proliferation assays, phospho-MAPK array, and mouse xenograft models. Here we demonstrated that TESC is overexpressed in colorectal cancer (CRC), but was not expressed in normal mucosa and premalignant dysplastic lesions. Furthermore, serum TESC levels were elevated in patients with CRC. Knockdown of TESC inhibited the Akt-dependent NF-κB pathway and decreased cell survival in vitro. Depletion of TESC reduced tumor growth in a CRC xenograft model. Thus, TESC is a potential diagnostic marker and oncotarget in colorectal cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human TESC
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tesc
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Serine, European Pharmacopoeia (EP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)