Merck
  • Glycosylation of immunoglobulin G determines osteoclast differentiation and bone loss.
Glycosylation of immunoglobulin G determines osteoclast differentiation and bone loss.
Nature communications (2015-04-01)
Ulrike Harre, Stefanie C Lang, René Pfeifle, Yoann Rombouts, Sabine Frühbeißer, Khaled Amara, Holger Bang, Anja Lux, Carolien A Koeleman, Wolfgang Baum, Katharina Dietel, Franziska Gröhn, Vivianne Malmström, Lars Klareskog, Gerhard Krönke, Roland Kocijan, Falk Nimmerjahn, René E M Toes, Martin Herrmann, Hans Ulrich Scherer, Georg Schett
ABSTRACT
Immunglobulin G (IgG) sialylation represents a key checkpoint that determines the engagement of pro- or anti-inflammatory Fcγ receptors (FcγR) and the direction of the immune response. Whether IgG sialylation influences osteoclast differentiation and subsequently bone architecture has not been determined yet, but may represent an important link between immune activation and bone loss. Here we demonstrate that desialylated, but not sialylated, immune complexes enhance osteoclastogenesis in vitro and in vivo. Furthermore, we find that the Fc sialylation state of random IgG and specific IgG autoantibodies determines bone architecture in patients with rheumatoid arthritis. In accordance with these findings, mice treated with the sialic acid precursor N-acetylmannosamine (ManNAc), which results in increased IgG sialylation, are less susceptible to inflammatory bone loss. Taken together, our findings provide a novel mechanism by which immune responses influence the human skeleton and an innovative treatment approach to inhibit immune-mediated bone loss.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Galactose, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99.0% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Millipore
D-(+)-Mannose, ≥99%, suitable for microbiology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SKU
Pack Size
Availability
Price
Quantity
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
SAFC
MOPS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
SKU
Pack Size
Availability
Price
Quantity
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)