Merck
  • Augmented EPR effect by photo-triggered tumor vascular treatment improved therapeutic efficacy of liposomal paclitaxel in mice bearing tumors with low permeable vasculature.
Augmented EPR effect by photo-triggered tumor vascular treatment improved therapeutic efficacy of liposomal paclitaxel in mice bearing tumors with low permeable vasculature.
Journal of controlled release : official journal of the Controlled Release Society (2015-01-03)
Tomoya Araki, Ken-ichi Ogawara, Haruka Suzuki, Rie Kawai, Taka-ichi Watanabe, Tsutomu Ono, Kazutaka Higaki
ABSTRACT
The effects of photo-triggered tumor vascular treatment (PVT) on the structural and functional properties of tumor vasculature were assessed in Colon-26 (C26) and B16/BL6 (B16) tumor-bearing mice. Furthermore, anti-tumor efficacy of subsequently injected PEG liposomal paclitaxel (PL-PTX) was also evaluated. As a photosensitizer, a hydrophobic porphyrin derivative was used and formulated in polymeric nanoparticle composed of polyethylene glycol-block-polylactic acid to avoid its non-specific in vivo disposition. In the mice bearing C26 with high permeable vasculature, the prominent anti-tumor activity was confirmed by PVT alone, but the subsequently injected PL-PTX did not show any additive effect. PVT itself initially induced apoptotic cell death of tumor vascular endothelial cells and platelet aggregation, which would have subsequently induced apoptosis of C26 tumor cells surrounding the vasculature. On the other hand, in the mice bearing B16 with low permeable vasculature, PVT enhanced the anti-tumor activity of subsequently injected PL-PTX, which would be attributed to the tumor disposition amount and area of PEG liposomes enhanced by PVT. These results clearly indicated that the treatment would have made it possible to provide more efficient extravasation of PL-PTX, leading to its more potent anti-tumor effect.
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Tin(II) 2-ethylhexanoate, 92.5-100.0%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
Cholesterol, Plant-Derived, SyntheChol®