Merck
The male-specific factor Sry harbors an oncogenic function.
Oncogene (2013-07-31)
S Murakami, S Chishima, H Uemoto, E Sakamoto, T Sato, N Kurabe, Y Kawasaki, T Shibata, H Akiyama, F Tashiro
ABSTRACT
Sgf29, a component of the SPT-ADA-GCN5 acetyltransferase (SAGA) complex, binds H3K4me2/3 marks and leads to histone H3 acetylation. Previously, we found that downregulation of Sgf29 suppresses c-Myc-mediated malignant transformation. Nonetheless, the upstream regulator of the Sgf29 gene is not yet known. Here, we report that Sry (sex-determining region Y), an HMG (high-mobility group) domain containing transcription factor, directly upregulates Sgf29 gene expression. Sry expression was deregulated in two out of the four tested male rodent hepatocellular carcinoma (rHCC) cell lines. Luciferase reporter and chromatin immunoprecipitation assays indicated that Sry could bind HMG-boxes in the proximal promoter region of the Sgf29 gene. Knockdown of Sry robustly lowered anchorage-independent growth, invasiveness and tumorigenicity of rHCC cells, whereas ectopic expression of Sry conferred more malignant properties. Thus, these data show that Sry is involved in male-specific malignant conversion of rHCCs via Sgf29 upregulation.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)