Merck
  • S-777469, a novel cannabinoid type 2 receptor agonist, suppresses itch-associated scratching behavior in rodents through inhibition of itch signal transmission.
S-777469, a novel cannabinoid type 2 receptor agonist, suppresses itch-associated scratching behavior in rodents through inhibition of itch signal transmission.
Pharmacology (2015-02-28)
Takayo Haruna, Masahiko Soga, Yasuhide Morioka, Ichiro Hikita, Kinichi Imura, Yoko Furue, Mina Yamamoto, Chihiro Imura, Minoru Ikeda, Akira Yamauchi, Masashi Deguchi, Michitaka Shichijo, Akinori Arimura, Kiyoshi Yasui
ABSTRACT
We have previously reported that S-777469 [1-([6-ethyl-1-(4-fluorobenzyl)-5-methyl-2-oxo-1,2-dihydropyridine-3-carbonyl]amino)-cyclohexanecarboxylic acid], a novel cannabinoid type 2 receptor (CB2) agonist, significantly suppressed compound 48/80-induced scratching behavior in mice in a dose-dependent manner when it was administered orally. Here, we demonstrated that the inhibitory effects of S-777469 on compound 48/80-induced scratching behavior are reversed by pretreatment with SR144528, a CB2-selective antagonist. In addition, we investigated the effects of S-777469 on itch-associated scratching behavior induced by several pruritogenic agents in mice and rats. S-777469 significantly suppressed scratching behavior induced by histamine or substance P in mice or by serotonin in rats. In contrast, the H1-antihistamine fexofenadine clearly inhibited histamine-induced scratching behavior but did not affect scratching behavior induced by substance P or serotonin. Moreover, S-777469 significantly inhibited histamine-induced peripheral nerve firing in mice. In conclusion, these results suggest that S-777469 produces its antipruritic effects by inhibiting itch signal transmission through CB2 agonism.
Sigma-Aldrich
Histamine, ≥97.0%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Histamine, analytical standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fexofenadine hydrochloride, >98% (HPLC)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Fexofenadine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Fexofenadine hydrochloride, United States Pharmacopeia (USP) Reference Standard