Merck
Alveolar-capillary adaptation to chronic hypoxia in the fatty lung.
Acta physiologica (Oxford, England) (2014-11-05)
C Yilmaz, P Ravikumar, D Gyawali, R Iyer, R H Unger, C C W Hsia
ABSTRACT
Obese diabetic (ZDF fa/fa) rats with genetic leptin resistance suffer chronic lipotoxicity associated with age-related lung restriction and abnormal alveolar ultrastructure. We hypothesized that these abnormalities impair adaptation to ambient hypoxia. Male fa/fa and lean (+/+) ZDF rats (4-months old) were exposed to 21 or 13% O2 for 3 weeks. Lung function was measured under anaesthesia. Lung tissue was assayed for DNA damage and ultrastructure measured by morphometry. In normoxia, lung volume, compliance and diffusing capacity were lower, while blood flow was higher in fa/fa than +/+ rats. In hypoxia, fa/fa animals lost more weight, circulating hematocrit rose higher, and lung volume failed to increase compared to +/+. In fa/fa, the hypoxia-induced increase in post-mortem lung volume was attenuated (19%) vs. +/+ (39%). Alveolar ducts were 35% smaller in normoxia but enlarged twofold more in hypoxia compared to +/+. Hypoxia induced broad increases (90-100%) in the volumes and surface areas of alveolar septal components in +/+ lungs; these increases were moderately attenuated in fa/fa lungs (58-75%), especially that of type II epithelium volume (16 vs. 61% in +/+). In fa/fa compared to +/+ lungs, oxidative DNA damage was greater with increased hypoxia induced efflux of alveolar macrophages. Harmonic mean thickness of the diffusion barrier was higher, indicating higher structural resistance to gas transfer. Chronic lipotoxicity impaired hypoxia-induced lung expansion and compensatory alveolar growth with disproportionate effect on resident alveolar progenitor cells. The moderate structural impairment was offset by physiological adaptation primarily via a higher hematocrit.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%