Merck
  • Antagonistic effects of indoloquinazoline alkaloids on antimycobacterial activity of evocarpine.
Antagonistic effects of indoloquinazoline alkaloids on antimycobacterial activity of evocarpine.
Journal of applied microbiology (2015-01-22)
C Hochfellner, D Evangelopoulos, M Zloh, A Wube, J D Guzman, T D McHugh, O Kunert, S Bhakta, F Bucar
ABSTRACT
The interaction of quinolone and indoloquinazoline alkaloids concerning their antimycobacterial activity was studied. The antimycobacterial and modulating activity of evodiamine (1), rutaecarpine (2) and evocarpine (3) was tested on mycobacteria including three multidrug-resistant (MDR) clinical isolates of Mycobacterium tuberculosis. Antagonistic effects were concluded from fractional inhibitory concentration (FICI) values. Interaction energies of the compounds were calculated using GLUE docking module implemented in GRID. 1 and 2 exhibited weak inhibition of rapidly growing mycobacteria, however, 1 was active against Myco. tuberculosis H37Rv (MIC = 10 mg l(-1) ) while 2 was inactive. Both 1 and 2 showed a marked antagonistic effect on the susceptibility of different mycobacterial strains to 3 giving FICI values between 5 and 9. The interaction energies between compounds 1 and 2 with compound 3 suggested the possibility of complex formation in solution. Indoloquinazoline alkaloids markedly reduce the antimycobacterial effect of the quinolone alkaloid evocarpine. Complex formation may play a role in the attenuation of its antimycobacterial activity. This study gives a striking example of antagonism between compounds present in the same plant extract which should be considered in natural product based screening projects.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Oleic acid, natural, FCC
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerol, analytical standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oleic acid, Selectophore, ≥99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture