Merck
  • Determination of nitrofuran and chloramphenicol residues by high resolution mass spectrometry versus tandem quadrupole mass spectrometry.
Determination of nitrofuran and chloramphenicol residues by high resolution mass spectrometry versus tandem quadrupole mass spectrometry.
Analytica chimica acta (2015-02-16)
A Kaufmann, P Butcher, K Maden, S Walker, M Widmer
ABSTRACT
An ultra-high performance liquid chromatography based method, coupled to high resolution mass spectrometry (UHPLC-HRMS), was developed to permit the detection and quantification of various nitrofuran and chloramphenicol residues in a number of animal based food products. This method is based on the hydrolysis of covalently bound metabolites and derivatization with 2-nitrobenzaldehyde. Clean-up is achieved by a liquid/liquid and a reversed phase/solid phase extraction. Not only are the four conventional nitrofurans (nitrofurantoin, furazolidone, nitrofurazone and furaltadone) detected, but also nifursol, nitrovin and nifuroxazide. Furthermore, an underivatizable nitrofuran (nifurpirinol) and another banned drug (chloramphenicol) can be quantified as well. The compounds are detected in the form of their precursor ions, [M+H](+) and [M-H](-), respectively. The mass resolving power of 70,000 FWHM, and the applied mass window ensure sufficient selectivity and sensitivity. Confirmation is obtained by monitoring the HRMS resolved product ions which were derived from the unit-mass resolved precursor ions. The multiplexing capability of the utilized Orbitrap instrument provides not only highly selective, but also sensitive confirmatory signals. This method has been validated according to the CD 2002/657/EC for the following matrices: muscle, liver, kidney, fish, honey, eggs and milk.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Nifuroxazide, VETRANAL®, analytical standard
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Nifuroxazide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material