Merck
  • Immunochemical detection of cytochrome P450 enzymes in small intestine microsomes of male and female untreated juvenile cynomolgus monkeys.
Immunochemical detection of cytochrome P450 enzymes in small intestine microsomes of male and female untreated juvenile cynomolgus monkeys.
Xenobiotica; the fate of foreign compounds in biological systems (2014-03-07)
Shotaro Uehara, Norie Murayama, Yasuharu Nakanishi, Chika Nakamura, Takanori Hashizume, Darryl C Zeldin, Hiroshi Yamazaki, Yasuhiro Uno
ABSTRACT
The expression of small intestinal cytochromes P450 (P450s) has not been systematically measured in cynomolgus monkeys, which are widely used in preclinical drug studies to predict pharmacokinetics and toxicity in humans: therefore, P450 content of small intestine was quantified in 35 cynomolgus monkeys by immunoblotting using 11 selective antibodies. CYP2D, CYP2J2, CYP3A4 and CYP3A5 were detected in all 35 animals, while CYP1A and CYP2C9/19 were detected in 31 and 17 animals, respectively. CYP2C9 and CYP2C19 were detected with the same antibody. CYP1D, CYP2A, CYP2B6, CYP2C76 and CYP2E1 were not detected in any of the 35 animals examined. On analysis of pooled microsomes (35 animals), CYP3A (3A4+3A5) was most abundant (79% of total immunoquantified CYP1-3 proteins), followed by CYP2J2 (13%), CYP2C9/19 (4%), CYP1A (3%) and CYP2D (0.4%). On the analysis of individual microsome samples, each P450 content varied 2-to-6-fold between animals, and no sex differences were observed in any P450 content. These findings should help to increase the understanding of drug metabolism, especially the first-pass effect, in cynomolgus monkey small intestines.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Acrylamide, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%