Merck
  • Poly-ADP-ribosylation of HMGB1 regulates TNFSF10/TRAIL resistance through autophagy.
Poly-ADP-ribosylation of HMGB1 regulates TNFSF10/TRAIL resistance through autophagy.
Autophagy (2015-01-22)
Minghua Yang, Liying Liu, Min Xie, Xiaofang Sun, Yan Yu, Rui Kang, Liangchun Yang, Shan Zhu, Lizhi Cao, Daolin Tang
ABSTRACT
Both apoptosis ("self-killing") and autophagy ("self-eating") are evolutionarily conserved processes, and their crosstalk influences anticancer drug sensitivity and cell death. However, the underlying mechanism remains unclear. Here, we demonstrated that HMGB1 (high mobility group box 1), normally a nuclear protein, is a crucial regulator of TNFSF10/TRAIL (tumor necrosis factor [ligand] superfamily, member 10)-induced cancer cell death. Activation of PARP1 (poly [ADP-ribose] polymerase 1) was required for TNFSF10-induced ADP-ribosylation of HMGB1 in cancer cells. Moreover, pharmacological inhibition of PARP1 activity or knockdown of PARP1 gene expression significantly inhibited TNFSF10-induced HMGB1 cytoplasmic translocation and subsequent HMGB1-BECN1 complex formation. Furthermore, suppression of the PARP1-HMGB1 pathway diminished autophagy, increased apoptosis, and enhanced the anticancer activity of TNFSF10 in vitro and in a subcutaneous tumor model. These results indicate that PARP1 acts as a prominent upstream regulator of HMGB1-mediated autophagy and maintains a homeostatic balance between apoptosis and autophagy, which provides new insight into the mechanism of TNFSF10 resistance.
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human FADD
Sigma-Aldrich
MISSION® esiRNA, targeting human HMGB1
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
PJ-34 hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human PARP1
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CASP8
Sigma-Aldrich
TRAIL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE and HPLC), lyophilized powder