Merck
  • Effects of hydrophobic and ionic interactions on glycation of casein during Maillard reaction.
Effects of hydrophobic and ionic interactions on glycation of casein during Maillard reaction.
Journal of agricultural and food chemistry (2014-11-05)
H Gül Akıllıoğlu, Vural Gökmen
ABSTRACT
This study aimed to investigate the effects of hydrophobic and ionic interactions on glycation of native and high-shear treated casein during heating. Casein-epicatechin and casein-calcium complexes were formed and glycated with glucose at different temperatures ranging from 70 to 150 °C in solution and dry states. Furosine, acid derivative of N-ε-fructoselysine (FL), and N-ε-carboxymethyl lysine (CML) were measured as indicators of early and advanced glycation, respectively. CML concentrations of casein-epicatechin and casein-calcium complexes heated in solution were significantly lower as compared to the control (p < 0.05). For instance, 182 ± 9.78 μg/g of CML formed in the control, while CML concentrations were 136 ± 10.7 and 101 ± 7.37 μg/g in casein-epicatechin and casein-calcium complexes, respectively, heated at 150 °C in the solution state. Treatment by high shear microfluidization further decreased the CML formed during heating at 70 °C in dry state. The results suggest that interactions with epicatechin molecule and calcium ion could be a useful strategy to limit advanced glycation of casein under certain conditions.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%