Merck
  • A colourimetric sensor for the simultaneous determination of oxidative status and antioxidant activity on the same membrane: N,N-dimethyl-p-phenylene diamine (DMPD) on Nafion.
A colourimetric sensor for the simultaneous determination of oxidative status and antioxidant activity on the same membrane: N,N-dimethyl-p-phenylene diamine (DMPD) on Nafion.
Analytica chimica acta (2015-03-04)
Sema Demirci Çekiç, Asli Neslihan Avan, Seda Uzunboy, Reşat Apak
ABSTRACT
A colourimetric sensor capable of simultaneously measuring oxidative status (OS) in terms of the hazard produced by reactive oxygen species (ROS) and antioxidant activity (AOA) in regard to ROS-scavenging ability of antioxidant compounds was developed. The coloured cationic semi-quinone derivatives, caused by ROS oxidative degradation of N,N-dimethyl-p-phenylene diamine hydrochloride (DMPD) in pH 5.7 acetate-buffered medium, were formed in solution and immobilized on a perfluorosulfonate-based Nafion membrane. ROS, namely hydroxyl (·OH) and superoxide (O2(·-)) radicals, were produced by Fenton/UV and xanthine/xanthine oxidase methods, respectively. The pink-coloured, (+)-charged chromophore (referred to as DMPD-quinone or DMPDQ), resulting from the reaction between DMPD and ROS, could be completely retained on the solid membrane sensor by electrostatic interaction with the anionic sulfonate groups of Nafion. After equilibration, the Nafion membrane surface was homogeneously coloured enabling an absorbance measurement at 514 nm, while the aqueous phase completely lacked colour. Antioxidants, when present, caused an absorbance decrease on the membrane due to their ROS scavenging action, giving rise to less DMPDQ production. The absorbance decrease on the sensor was linearly dependent on antioxidant concentration over a reasonable concentration range, enabling the simultaneous determination of OS and AOA-against ROS. The proposed antioxidant sensing method was tested in synthetic and real antioxidant mixtures, and validated against standard antioxidant capacity assays (i.e. ABTS and CUPRAC) for a variety of polyphenolic and antioxidant compounds. The dynamic linear ranges of antioxidants with the DMPD sensor in protection against hydroxyl and superoxide radicals generally varied within the micromolar to a few tens of micromolar concentration interval over one order-of-magnitude. Choosing three representative compounds in the high (epigallocatechin gallate), medium (quercetin) and low (p-coumaric acid) molar absorptivity range, the detection limits ranged within the concentration intervals of 0.2-0.9 μM, 0.3-0.8 μM, and 4-14 μM, respectively, depending on the radical scavenged.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-Naringenin, ≥95%