Merck
  • Oxidative DNA damage and disturbance of antioxidant capacity by alternariol in Caco-2 cells.
Oxidative DNA damage and disturbance of antioxidant capacity by alternariol in Caco-2 cells.
Toxicology letters (2015-04-02)
Celia Fernández-Blanco, Guillermina Font, Maria-Jose Ruiz
ABSTRACT
Oxidative stress occurs as a consequence of an imbalance between the prooxidant/antioxidant systems, causing an increase of intracellular generation of reactive oxygen species. Alternariol (AOH), a mycotoxin produced by Alternaria sp. can alter the action of glutathione (GSH) and the enzymes involved in the redox system, causing damage to cellular macromolecules such as DNA. The aims of this work were to determine the induction of oxidative stress by the antioxidant defenses imbalance in relation to glutathione (GSH), glutathione reductase (GR), glutathione transferase (GST), glutathione peroxidase (GPx) levels and DNA damage in Caco-2 cells derived from adenocarcinoma human colon. Oxidative stress by AOH was confirmed by alteration of GSH levels and the antioxidant defense system after 15, 30 and 60 μM AOH exposure during 24h. GSH levels significantly decreased by 43% after treatment with 60 μM AOH compared to the control. The activity of GPx and GR was reduced by 30% and 23%, respectively after 60 μM AOH. The GST activity was significantly increased (approximately 22%) with 30 μM AOH, while 60 μM AOH decreased it by 30% in comparison to the control. Analysis of DNA damage was performed using the Comet assay after 24h, where the % of DNA in tail increased from 70% to 85% compared the control.
Product Description
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications