Merck
  • Acromegaly is protective for periodontal tissue - advanced chronic periodontitis is rare in acromegalics.
Acromegaly is protective for periodontal tissue - advanced chronic periodontitis is rare in acromegalics.
Bratislavske lekarske listy (2014-10-17)
S Bascil, H Serinsoz, N B Tutuncu
ABSTRACT
Acromegaly is a complex syndrome which influences the connective tissue and bone metabolism. Although the effects of excess growth hormone on bone tissue are well known, its effect on periodontal tissue is an enigma. A total of 23 acromegalic patients (mean age 48.13 ± 9.4 years) were included randomly in to this cross-sectional study. The control group consisted of 60 healthy (mean age 50.3 ± 8.5) individuals. All participants were examined by the same periodontist to evaluate periodontal disease status. We studied a group of 23 acromegalic patients (11 females, 12 males) with estimated duration of disease from minimum of one year to 10 years (mean 4.52 ± 2.9 years). The frequency of advanced chronic periodontitis in acromegalics was significantly lower than in the healthy group (p = 0.002). Advanced periodontitis was 10 times more frequent in healthy population than in the acromegalics. Advanced chronic periodontitis is 10 times less frequent in acromegalics than in healthy population. Acromegaly is a disease which seems to be protective for periodontal tissue (Tab. 4, Ref. 30).
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications