Merck
  • A novel approach for electromembrane extraction based on the use of silver nanometallic-decorated hollow fibers.
A novel approach for electromembrane extraction based on the use of silver nanometallic-decorated hollow fibers.
Analytica chimica acta (2014-10-11)
María Ramos-Payán, Rut Fernández-Torres, Juan Luis Pérez-Bernal, Manuel Callejón-Mochón, Miguel Ángel Bello-López
ABSTRACT
A novel approach based on the use of nanometallic-decorated hollow fibers to assist electromembrane extraction is proposed. Microporous polypropylene hollow fibers, on which nanometallic silver was deposited, have been used for the first time as liquid membrane support in electromembrane extraction (EME). Different methods for the generation/deposition of silver nanoparticles (AgNPs) were studied. The best results were obtained with chemical reduction of silver nitrate using NaBH4 in aqueous solution followed by direct deposition on the hollow fibers. The extraction performance of the new supports was compared with a previously developed EME procedure used for the extraction of selected non-steroidal anti-inflammatory drugs (NSAIDs), resulting in an increase in the extraction ratio by a factor of 1.2-2 with a 30% reduction in the extraction time. The new nanometallic-decorated supports open new possibilities for EME due to the singular properties of nanometallic particles, including chemical fiber functionalization.
Product Description
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
VenPure® SF, powder
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline