Merck
  • A reproducible immunopotency assay to measure mesenchymal stromal cell-mediated T-cell suppression.
A reproducible immunopotency assay to measure mesenchymal stromal cell-mediated T-cell suppression.
Cytotherapy (2014-12-03)
Debra D Bloom, John M Centanni, Neehar Bhatia, Carol A Emler, Diana Drier, Glen E Leverson, David H McKenna, Adrian P Gee, Robert Lindblad, Derek J Hei, Peiman Hematti
ABSTRACT
The T-cell suppressive property of bone marrow-derived mesenchymal stromal cells (MSCs) has been considered a major mode of action and basis for their utilization in a number of human clinical trials. However, there is no well-established reproducible assay to measure MSC-mediated T-cell suppression. At the University of Wisconsin-Madison Production Assistance for Cellular Therapy (PACT) Center, we developed an in vitro quality control T-cell suppression immunopotency assay (IPA) that uses anti-CD3 and anti-CD28 antibodies to stimulate T-cell proliferation. We measured MSC-induced suppression of CD4+ T-cell proliferation at various effector-to-target cell ratios with the use of defined peripheral blood mononuclear cells and in parallel compared with a reference standard MSC product. We calculated an IPA value for suppression of CD4+ T cells for each MSC product. Eleven MSC products generated at three independent PACT centers were evaluated for cell surface phenotypic markers and T-cell suppressive properties. Flow cytometry results demonstrated typical MSC cell surface marker profiles. There was significant variability in the level of suppression of T-cell proliferation, with immunopotency assay values ranging from 27% to 88%. However, MSC suppression did not correlate with human leukocyte antigen-DR expression. We have developed a reproducible immunopotency assay to measure allogeneic MSC-mediated suppression of CD4+ T cells. Additional studies may be warranted to determine how these in vitro assay results may correlate with other immunomodulatory properties of MSCs, in addition to evaluating the ability of this assay to predict in vivo efficacy.
Product Description
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Aphidicolin, analytical standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar