Merck
  • Investigation on performance of zirconia and magnesia-zirconia stationary phases in hydrophilic interaction chromatography.
Investigation on performance of zirconia and magnesia-zirconia stationary phases in hydrophilic interaction chromatography.
Talanta (2014-08-17)
Qing Wang, Jing Li, Xin Yang, Li Xu, Zhi-guo Shi, Lan-Ying Xu
ABSTRACT
In the current study, zirconia (ZrO2) and its composite, magnesia-zirconia (MgO-ZrO2), were prepared as the hydrophilic interaction chromatographic (HILIC) stationary phases (SPs). Different experimental variables including water content, pH and buffer concentration in the mobile phase (MP) as well as column temperature were systematically studied to permit an in-depth understanding of the chromatographic properties of the mentioned SPs and to explore the retention mechanism further on. The results were compared with a native SiO2 column. Adsorption was demonstrated as the main retention mechanism on the two ZrO2-based SPs. The transferring of the analytes from the MP to the ZrO2-based SPs was endothermic and high column temperature would facilitate the retention. In addition, the MgO-ZrO2 SP exhibited superior resolution, column efficiency as well as stronger retention in comparison to the bare ZrO2 SP, which demonstrated that the introduction of MgO could improve the structure and properties of the material. In conclusion, MgO-ZrO2 was a promising material for HILIC applications.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Supelco
Sulfamerazine, VETRANAL®, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm