Merck
  • Pharmacokinetics and tissue distribution of five bufadienolides from the Shexiang Baoxin Pill following oral administration to mice.
Pharmacokinetics and tissue distribution of five bufadienolides from the Shexiang Baoxin Pill following oral administration to mice.
Journal of ethnopharmacology (2014-09-10)
Huimei Huang, Yongge Yang, Chao Lv, Wanlin Chang, Chengcheng Peng, Shuping Wang, Guangbo Ge, Lin Han, Weidong Zhang, Runhui Liu
ABSTRACT
Shexiang Baoxin Pill (SBP) is a well-known composite formula of traditional Chinese medicine (TCM), widely used to treat cardiovascular diseases such as angina pectoris and myocardial infarction. Bufadienolides are major active compounds of Venenum Bufonis, which is one of the seven materiamedicas that comprise the Shexiang Baoxin Pill. Previous pharmacokinetics studies of bufadienolides have typically used a single medicinal material delivered to rats. In this study, we have chosen the mouse, a more proper animal model than the rat, to investigate the in vivo pharmacokinetics and tissue distribution of bufadienolides from the Shexiang Baoxin Pill. The concentrations of bufadienolides in plasma and tissues were identified using high performance liquid chromatography-tandem mass spectrometry (HPLC-ESI-MS/MS). The samples were prepared by liquid-liquid extraction with ethyl acetate, and the separation of bufadienolides was achieved using an ACQUITY HSS T3 column by gradient elution using water (containing 0.1% formic acid) and acetonitrile as the mobile phase at a flow rate of 0.3 mL/min. The pharmacokinetic parameters were determined using non-compartmental analysis. The results showed that the five bufadienolides were rapidly absorbed and distributed into the body. The pharmacokinetic curve showed double peaks after oral administration. The major tissue depots for resibufogenin, bufalin, and bufotalin in mice were the intestines, lung and kidney, whereas the major tissue depots of gamabufotalin and arenobufagin were the intestines, liver and kidney. The information gained from this research provides a meaningful insight for the clinical applications of the Shexiang Baoxin Pill.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%