Merck
  • Concentrations of venlafaxine and its main metabolite O-desmethylvenlafaxine during pregnancy.
Concentrations of venlafaxine and its main metabolite O-desmethylvenlafaxine during pregnancy.
Journal of clinical pharmacy and therapeutics (2014-07-06)
P G J ter Horst, K H M Larmené-Beld, J Bosman, E L van der Veen, A Wieringa, J P Smit
ABSTRACT
Depression during pregnancy is common and includes risks for mother and child. Pharmacokinetics of venlafaxine may be changed during pregnancy. This study aimed to describe changes in metabolic ratios and concentrations of venlafaxine and its main metabolite O-desmethylvenlafaxine during and after pregnancy. To study this, we used data from our study of compliance to Antidepressants During Pregnancy (the ADAP study) to investigate the course of venlafaxine and O-desmethylvenlafaxine concentrations during pregnancy and in the period post-partum. We found that the venlafaxine concentration significantly changed during pregnancy when compared to the post-partum period (P = 0·028). The median concentration of venlafaxine in the first trimester was 98·9% (54·2-292·0%), the second 100·0% (46·5-264·0%) and the third trimester 87·0% (61·5-217·2%). We did not found differences in O-desmethylvenlafaxine concentrations in the different trimesters of pregnancy compared with the post-partum period, P = 0·565. Also the ratio of O-desmethylvenlafaxine/venlafaxine concentrations increased significantly from 76·9% (range 32·8-142·0%) in the first trimester to 196·7% (range 83·3-427·6%) in the third trimester compared with the post-partum period, P = 0·004. Further, three of seven patients had concentrations below the therapeutic reference range (100-400 μg/L) in any period of pregnancy, whereas no one had subtherapeutic concentrations in the post-partum period. Venlafaxine concentrations decreases during pregnancy, and the ratio of the concentrations of O-desmethylvenlafaxine/venlafaxine increases during pregnancy. Pregnant women using venlafaxine are at risk for subtherapeutic concentrations, therefore routine monitoring of concentrations venlafaxine and O-desmethylvenlafaxine is recommendable during pregnancy.
Sigma-Aldrich
Butyl methyl ether, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%