Merck
  • β-Hemolytic streptococcal throat carriage and tonsillopharyngitis: a cross-sectional prevalence study in Gabon, Central Africa.
β-Hemolytic streptococcal throat carriage and tonsillopharyngitis: a cross-sectional prevalence study in Gabon, Central Africa.
Infection (2014-11-29)
Sabine Bélard, Nicole Toepfner, Benjamin Arnold, Abraham Sunday Alabi, Reinhard Berner
ABSTRACT
Group A streptococcus (GAS) and possibly other β-hemolytic streptococci (BHS) account for a considerable morbidity and mortality burden in African populations; however, disproportionately little is known about the epidemiology of BHS in sub-Saharan Africa. This study assessed the prevalence of GAS, group G streptococcus (GGS) and group C streptococcus (GCS) carriage and tonsillopharyngitis in a Central African population. A prospective cross-sectional study was performed to assess the prevalence of and risk factors for BHS carrier status and tonsillopharyngitis in children and adults in Gabon. The overall BHS carrier prevalence was 135/1,005 (13.4%); carrier prevalence of GAS, GGS, and GCS was 58/1,005 (5.8%), 50/1,005 (5.0%), and 32/1,005 (3.2%), respectively. Streptococcal carriage was associated with school and pre-school age (adjusted OR 2.65, 95% CI 1.62-4.36, p = 0.0001 and 1.90, 95% CI 1.14-3.17, p = 0.0141, respectively). Participants residing in urban areas were less likely carriers (OR 0.52, p = 0.0001). The point-prevalence of BHS-positive tonsillopharyngitis was 1.0% (9/1,014) and 15.0% (6/40) in school children with sore throat. Non-GAS exceeded GAS throat carriage and tonsillopharyngitis suggesting a yet underestimated role of non-GAS streptococci in BHS diseases.
Product Description
Supelco
4,4′-DDT, PESTANAL®, analytical standard
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%