Merck
  • +TIP EB1 downregulates paclitaxel‑induced proliferation inhibition and apoptosis in breast cancer cells through inhibition of paclitaxel binding on microtubules.
+TIP EB1 downregulates paclitaxel‑induced proliferation inhibition and apoptosis in breast cancer cells through inhibition of paclitaxel binding on microtubules.
International journal of oncology (2014-10-14)
Geethu Emily Thomas, Jamuna S Sreeja, K K Gireesh, Hindol Gupta, Tapas K Manna
ABSTRACT
Microtubule plus‑end‑binding protein (+TIP) EB1 has been shown to be upregulated in breast cancer cells and promote breast tumor growth in vivo. However, its effect on the cellular actions of microtubule‑targeted drugs in breast cancer cells has remained poorly understood. By using cellular and biochemical assays, we demonstrate that EB1 plays a critical role in regulating the sensitivity of breast cancer cells to anti‑microtubule drug, paclitaxel (PTX). Cell viability assays revealed that EB1 expression in the breast cancer cell lines correlated with the reduction of their sensitivity to PTX. Knockdown of EB1 by enzymatically‑prepared siRNA pools (esiRNAs) increased PTX‑induced cytotoxicity and sensitized cells to PTX‑induced apoptosis in three breast cancer cell lines, MCF‑7, MDA MB‑231 and T47D. Apoptosis was associated with activation of caspase‑9 and an increase in the cleavage of poly(ADP‑ribose) polymerase (PARP). p53 and Bax were upregulated and Bcl2 was downregulated in the EB1‑depleted PTX‑treated MCF‑7 cells, indicating that the apoptosis occurs via a p53‑dependent pathway. Following its upregulation, the nuclear accumulation of p53 and its association with cellular microtubules were increased. EB1 depletion increased PTX‑induced microtubule bundling in the interphase cells and induced formation of multiple spindle foci with abnormally elongated spindles in the mitotic MCF‑7 cells, indicating that loss of EB1 promotes PTX‑induced stabilization of microtubules. EB1 inhibited PTX‑induced microtubule polymerization and diminished PTX binding to microtubules in vitro, suggesting that it modulates the binding sites of PTX at the growing microtubule ends. Results demonstrate that EB1 downregulates inhibition of PTX‑induced proliferation and apoptosis in breast cancer cells through a mechanism in which it impairs PTX‑mediated stabilization of microtubule polymerization and inhibits PTX binding on microtubules.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder