Merck
  • In depth analysis of apoptosis induced by silica coated manganese oxide nanoparticles in vitro.
In depth analysis of apoptosis induced by silica coated manganese oxide nanoparticles in vitro.
Journal of hazardous materials (2014-12-03)
Chao Yu, Zhiguo Zhou, Jun Wang, Jin Sun, Wei Liu, Yanan Sun, Bin Kong, Hong Yang, Shiping Yang
ABSTRACT
Manganese oxide nanoparticles (MnO NPs) have been regarded as a new class of T1-positive contrast agents. The cytotoxicity of silica coated MnO NPs (MnO@SiO2 NPs) was investigated in human cervical carcinoma cells (HeLa) and mouse fibroblast cells (L929). The changes of cell viability, cell morphology, cellular oxidative stress, mitochondrial membrane potential and cell cycle induced by MnO@SiO2 NPs were evaluated. Compared to HeLa cells, L929 cells showed lower cell viability, more strongly response to oxidative stress and higher percentage in the G2/M phase of cell cycle. The appearance of sub-G1 peak, double staining with Annexin V-FITC/PI and the increase of Caspase-3 activity further confirmed apoptosis should be the major form of cell death. Moreover, the apoptotic pathway was clarified as follows. Firstly, reactive oxygen species (ROS) is generated induced by MnO@SiO2 NPs, then p53 is activated followed by an increase in the bax and a decrease in the bcl-2, ultimately leading to G2/M phase arrest, increasing the activity of caspase-3 and inducing apoptosis.
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Supelco
Cyclohexane, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O