Merck
  • Polyproline promotes tetramerization of recombinant human butyrylcholinesterase.
Polyproline promotes tetramerization of recombinant human butyrylcholinesterase.
The Biochemical journal (2014-06-12)
Marilynn A Larson, Oksana Lockridge, Steven H Hinrichs
ABSTRACT
Human BChE (butyrylcholinesterase) protects against the toxicity of organophosphorus nerve agents and pesticides. BChE purified from human plasma is limited and pathogen carry-over is a concern. Unlike the native BChE tetrameric complex with a residence time of days, rBChE (recombinant BChE) is produced predominantly as dimers and monomers that are cleared from the circulation within minutes. Assembly into tetramers requires incorporation of proline-rich peptides, a process that was thought to occur intracellularly. Our goal was to determine whether polyproline added to rBChE under cell-free conditions would promote tetramerization. Secreted rBChE was purified by procainamide affinity chromatography, and synthetic polyprolines (8-mer to 300-mer) were tested to determine their effect on tetramer assembly. These studies demonstrated that 90-98% of purified rBChE (65 μM) could be assembled into tetramers when incubated with synthetic 17-mer or 50-mer polyproline peptides (100 μM) for 1.5 h at 25°C. However, rBChE tetramerization was inefficient with smaller 8-mer polyproline peptides and larger 300-mer polyproline proteins. Collectively, these studies demonstrated that the eukaryotic cellular machinery is not required for assembly of active BChE into tetramers and that this process can occur in vitro with purified rBChE in the presence of peptides containing 15-50 consecutive proline residues.
Product Description
Supelco
L-Methionine sulfoximine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine sulfoximine, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material