Merck
  • Silica-based nanofibers for electrospun ultra-thin layer chromatography.
Silica-based nanofibers for electrospun ultra-thin layer chromatography.
Journal of chromatography. A (2014-09-15)
Toni E Newsome, Susan V Olesik
ABSTRACT
Nanofibrous silica-based stationary phases for electrospun ultra-thin layer chromatography (E-UTLC) are described. Nanofibers were produced by electrospinning a solution of silica nanoparticles dispersed in polyvinylpyrrolidone solutions to create composite silica/polymer nanofibers. Stationary phases were created from as-spun nanofibers, or the nanofibers were heated either to crosslink the polyvinylpyrrolidone or to calcine and selectively remove the polymer. As-spun, crosslinked, and calcined nanofibers with similar mat thicknesses (23-25 μm) were evaluated as stationary phases for E-UTLC separations of laser dyes and amino acids and compared to commercial silica TLC plates. As-spun nanofiber plates offered fast mobile phase velocities, but like other polymer-based nanofibers, separations were only compatible with techniques using nonsolvents of the polymer. Crosslinked nanofibers were not as limited in terms of chemical stability, but separations produced tailed spot shapes. No limitations in terms of mobile phases, analyte solvents, and visualization techniques were observed for calcined nanofibers. Highly efficient separations of amino acids were performed in 15 mm on calcined nanofiber plates, with observed plate heights as low as 8.6 μm, and plate numbers as large as 1400. Additional alignment of the nanofibers provided shorter analysis times but also larger spot widths. The extension of stationary phases to silica-based nanofibers vastly expands the range of mobile phases, analyte solvents, and visualization techniques which can be used for E-UTLC separations.
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
Acetic acid, analytical standard
Supelco
Acetone, analytical standard
Supelco
Heptane, analytical standard
Supelco
1-Propanol, analytical standard
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture