Merck
  • Determination of irinotecan and its metabolite SN-38 in rabbit plasma and tumors using a validated method of tandem mass spectrometry coupled with liquid chromatography.
Determination of irinotecan and its metabolite SN-38 in rabbit plasma and tumors using a validated method of tandem mass spectrometry coupled with liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-14)
Dae J Park, Jun H Won, A R Cho, Hye J Yun, Jeong H Heo, Tae H Hwhang, Dae H Lee, Woo M Kim
ABSTRACT
New tandem mass spectrometric method coupled with liquid chromatography (LC-MS/MS) has been developed to determine the total concentration of camptothecin derivatives (irinotecan and SN-38) regardless of inter-conversion phenomenon between carboxylate and lactone forms. At first, all sample solutions were acidified for 1h in order to completely convert CPT derivatives into their lactone forms and then CPT derivatives were extracted with organic solution containing diethyl ether and ethyl acetate (2:1, v/v) just after alkalization in the range pH 8.0-8.5 in acid-treated solutions. Analytes were separated on a reverse phase C18 column (150×2.1mm) and eluted isocratically with a mobile phase which consisted of acetonitrile-methanol-buffer (0.1% formic acid, 5mM ammonium formate) (3:4:3, v/v). CPT derivatives were monitored by tandem mass spectrometry in electrospay-positive ionization and multiple reaction mode programmed to the following transitions (m/z): '587.6→167.2' of CPT-11, '393.6→349.3' of SN-38 and '349.4→ 305.2' of CPT. The method was validated to have the proper linearity (r(2)>0.99) over the range of 5-1000ng/ml of CPT-11 and 1-250ng/ml of SN-38 with good accuracy (89.8-114.3%) and precision (less than 10%). In all stability tests, concentration of CPT-11 and SN-38 had been left in the acceptable range of 88.8-110.7% when sample solutions were acidified before determination of CPT derivatives. Newly developed LC-MS/MS method was suitable for the determination of CPT derivatives of both rabbit plasma and tumor tissues in the pharmacokinetic study.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl acetate