Merck
  • Fast determination of 22 sulfonamides from chicken breast muscle using core-shell nanoring amino-functionalized superparamagnetic molecularly imprinted polymer followed by liquid chromatography-tandem mass spectrometry.
Fast determination of 22 sulfonamides from chicken breast muscle using core-shell nanoring amino-functionalized superparamagnetic molecularly imprinted polymer followed by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-05-02)
Yong-Gang Zhao, Li-Xin Zhou, Sheng-Dong Pan, Ping-Ping Zhan, Xiao-Hong Chen, Mi-Cong Jin
ABSTRACT
A novel, simple and sensitive method was developed for the simultaneous determination of 22 sulfonamides (SAs) in chicken breast muscle by using the dispersive micro-solid-phase extraction (d-μ-SPE) procedure combined with ultra-fast liquid chromatography-tandem quadrupole mass spectrometry (UFLC-MS/MS). The excellent core-shell nanoring amino-functionalized superparamagnetic molecularly imprinted polymer (CS-NR-Mag-MIP) was used as sorbent, and the main factors affecting the extraction efficiency were investigated in detail. All target compounds showed good linearities in the tested range with correlation coefficients (r) higher than 0.9980. The mean recoveries were in the range of 85.0-112.2% at low, medium and high concentration levels (0.1, 1.0 and 5.0 ng/g). The intra-day and inter-day relative standard deviations (RSDs) were lower than 6.0% and 8.9%, respectively. The limits of quantification for the 22 SAs were between 0.013 and 0.099 ng/g. The developed d-μ-SPE-UFLC-MS/MS method had been successfully applied to the chicken breast muscle samples for food-safety risk monitoring in Zhejiang Province, China. The results showed sulfamethazine, sulfamethoxazole and sulfaquinoxaline in five out of three hundred collected samples were detected with concentrations of 0.418-2.28, 16.4 and 2.93 ng/g, respectively. It was confirmed that the CS-NR-Mag-MIP was a kind of highly effective d-μ-SPE materials used for the SAs analyses.
Product Description
Supelco
Sulfadoxin, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Sulfameter
Supelco
Sulfamethizole, analytical standard, ≥99% (HPLC)
Sigma-Aldrich
Sulfamonomethoxine
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard
Supelco
Sulfaguanidine, VETRANAL®, analytical standard
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfaguanidine
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfaquinoxaline, PESTANAL®, analytical standard
Supelco
Sulfapyridine melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethizole, VETRANAL®, analytical standard
USP
Sulfaquinoxaline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)