Merck
  • Transcriptomic profiling of human choriodecidua during term labor: inflammation as a key driver of labor.
Transcriptomic profiling of human choriodecidua during term labor: inflammation as a key driver of labor.
American journal of reproductive immunology (New York, N.Y. : 1989) (2014-10-07)
Gillian L Stephen, Sylvia Lui, Sarah A Hamilton, Clare L Tower, Lynda K Harris, Adam Stevens, Rebecca L Jones
ABSTRACT
Inflammation is a driver of labor in myometrium and cervix; however, the involvement of decidua is poorly defined. We have reported decidual leukocyte infiltration prior to and during labor; the regulators of these inflammatory processes are unknown. Choriodecidua RNA obtained after term labor or elective cesarean delivery was applied to Affymetrix GeneChips. Pathway analysis and gene validation were performed. Extensive inflammatory activation was identified in choriodecidua following labor, predominantly upregulation of genes regulating leukocyte trafficking and cytokine signalling. Genes governing cell fate, tissue remodelling, and translation were also altered. Upregulation of candidate genes (ICAM1, CXCR4, CD44, TLR4, SOCS3, BCL2A, and IDO) was confirmed. NFκB, STAT1&3, HMGB1, and miRNA-21, miRNA-46, miRNA-141, and miRNA-200 were predicted upstream regulators. This study confirms inflammatory processes are major players in labor events in choriodecidua, as in other gestational tissues. Suppressing uterine inflammation is likely to be critical for arresting premature labor.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%