Merck
  • Antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species.
Antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species.
International journal of pharmaceutics (2014-06-19)
Li Li, Hana Cho, Kwon Hyeok Yoon, Han Chang Kang, Kang Moo Huh
ABSTRACT
Poly(ethylene glycol)-b-poly(caprolactone) (PEG-b-PCL) micelles dually loaded with both pheophorbide a (PhA) as a photosensitizer and β-carotene (CAR) as a singlet oxygen ((1)O2) scavenger were designed to control photodynamic therapy (PDT) activity in cancer treatment. The CAR in the PhA/CAR micelles significantly diminished PhA-generated (1)O2 through direct (1)O2 scavenging, whereas the CAR molecules lost their (1)O2 scavenging activity when the PhA and CAR were spatially isolated by the disintegration of the PEG-b-PCL micelles. In cell-culture systems, light irradiation at a post-treatment time that corresponded to the presence of the micelles in the blood environment induced negligible phototoxicity, whereas light irradiation at a post-treatment time that corresponded to the presence of the micelles in the intracellular environment induced remarkable phototoxicity. In addition, a longer post-treatment time induced greater internalization of PhA/CAR micelles, which resulted in higher phototoxicity, suggesting an increase in photo killing activity against the tumor cells of interest. Thus, the co-loading of a (1)O2 generator and a (1)O2 scavenger into a single micelle is a potential strategy that may be useful in facilitating more accurate and reliable PDT with site-specific controllable production of singlet oxygen species for cancer treatment.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.