Merck
  • Development of a new cucumber reference material for pesticide residue analysis: feasibility study for material processing, homogeneity and stability assessment.
Development of a new cucumber reference material for pesticide residue analysis: feasibility study for material processing, homogeneity and stability assessment.
Analytical and bioanalytical chemistry (2015-01-30)
Susana Grimalt, Stefan Harbeck, Penka Shegunova, John Seghers, Berit Sejerøe-Olsen, Håkan Emteborg, Marta Dabrio
ABSTRACT
The feasibility of the production of a reference material for pesticide residue analysis in a cucumber matrix was investigated. Cucumber was spiked at 0.075 mg/kg with each of the 15 selected pesticides (acetamiprid, azoxystrobin, carbendazim, chlorpyrifos, cypermethrin, diazinon, (α + β)-endosulfan, fenitrothion, imazalil, imidacloprid, iprodione, malathion, methomyl, tebuconazole and thiabendazole) respectively. Three different strategies were considered for processing the material, based on the physicochemical properties of the vegetable and the target pesticides. As a result, a frozen spiked slurry of fresh cucumber, a spiked freeze-dried cucumber powder and a freeze-dried cucumber powder spiked by spraying the powder were studied. The effects of processing and aspects related to the reconstitution of the material were evaluated by monitoring the pesticide levels in the three materials. Two separate analytical methods based on LC-MS/MS and GC-MS/MS were developed and validated in-house. The spiked freeze-dried cucumber powder was selected as the most feasible material and more exhaustive studies on homogeneity and stability of the pesticide residues in the matrix were carried out. The results suggested that the between-unit homogeneity was satisfactory with a sample intake of dried material as low as 0.1 g. A 9-week isochronous stability study was undertaken at -20 °C, 4 °C and 18 °C, with -70 °C designated as the reference temperature. The pesticides tested exhibited adequate stability at -20 °C during the 9-week period as well as at -70 °C for a period of 18 months. These results constitute a good basis for the development of a new candidate reference material for selected pesticides in a cucumber matrix.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Malathion, certified reference material, TraceCERT®
Supelco
Chlorpyrifos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cyclohexane, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Fenitrothion, PESTANAL®, analytical standard
Supelco
Iprodione, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Chlorpyrifos, PESTANAL®, analytical standard
Supelco
Thiabendazole, PESTANAL®, analytical standard
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS