Merck
  • Novel pyrano and vinylphenol adducts of deoxyanthocyanidins in sorghum sourdough.
Novel pyrano and vinylphenol adducts of deoxyanthocyanidins in sorghum sourdough.
Journal of agricultural and food chemistry (2014-11-06)
Yunpeng Bai, Brandon Findlay, Alma Fernanda Sanchez Maldonado, Andreas Schieber, John C Vederas, Michael G Gänzle
ABSTRACT
This study determined the fate of deoxyanthocyanidins in sorghum sourdoughs. Sourdoughs prepared from the red sorghum variety Town were fermented with the caffeic acid-decarboxylating strains Lactobacillus plantarum FUA3171 and the decarboxylase negative L. casei FUA3166. Deoxyanthocyanidins were analyzed by liquid chromatography-tandem mass spectrometry (LC-MS/MS). Apigeninidin and methoxyapigeninidin were the major deoxyanthocyanidins prior to fermentation. During fermentation, novel deoxyanthocyanidins were formed. Purification by preparative LC, followed by NMR analysis and high-resolution MS identified two of the compounds as 6-deoxyanthocyanidin-vinylphenol and pyrano-3-deoxyanthocyanidin. To identify pathways for their formation, sorghum was fermented with single strains, L. plantarum or L. casei. 6-Deoxyanthocyanidin-vinylphenol and pyrano-3-deoxyanthocyanidin were formed only during fermentation with L. plantarum FUA3171, indicating a role of vinylphenol in their formation. Chemical synthesis confirmed that 6-deoxyanthocyanidin-vinylphenol and pyrano-3-deoxyanthocyanidin are formed from apigeninidin with vinylphenol but not with p-coumaric acid as reactants. In conclusion, the products of microbial decarboxylation of hydroxycinnamic acids convert apigeninidin and methoxyapigeninidin to pyrano-3-deoxyanthocyanidins and vinylphenol adducts.
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%