Merck
  • p63 regulates growth of esophageal squamous carcinoma cells via the Akt signaling pathway.
p63 regulates growth of esophageal squamous carcinoma cells via the Akt signaling pathway.
International journal of oncology (2014-04-11)
Shuai Ye, Kwang Bok Lee, Man Hee Park, Ju-Seog Lee, Soo Mi Kim
ABSTRACT
p63 is a member of the p53 protein family and plays a crucial role in epithelial development. p63 is expressed in many types of tumors including esophageal cancer; however, its function in cancer is controversial and its role in esophageal cancer has not been clearly established. In the present study, we aimed to identify the mechanisms by which p63 promotes proliferation of esophageal squamous carcinoma cells. Four human esophageal cancer cell lines (TE-8, TE-12, BE3 and OE33) were used in this study. We found that ΔNp63 was the predominantly expressed p63 isoform in esophageal squamous cancer cells. Silencing of p63 mRNA in the esopha-geal cancer cell lines TE-8 and TE-12 resulted in significant inhibition of cell proliferation in a dose-dependent manner. A colony formation assay also showed that colony formation by TE-8 and TE-12 cells was significantly inhibited by p63 siRNA. Furthermore, p63 siRNA significantly suppressed p-Akt and induced Akt expression in esophageal squamous carcinoma cell lines. On the other hand, overexpression of p63 in the esophageal cell lines BE3 and OE33 increased p-Akt expression. Silencing of p63 in TE-8 and TE-12 cell lines induced p53 and p27 expression and suppressed cyclin D1 and cyclin E1 expression, whereas overexpression of p63 in BE3 and OE33 cell lines resulted in decreased levels of p53 and p27 and increased levels of cyclin D1 and cyclin E1. Taken together, our results suggest that p63 may play a pivotal role in the progression of esophageal squamous carcinoma cells through regulation of the cell cycle via the Akt signaling pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human GAPDH
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tcp1
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gapdh
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human TCP1
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Trp63
Sigma-Aldrich
MISSION® esiRNA, targeting human TP63
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard