Merck
  • Anticoagulant mechanism and platelet deaggregation property of a non-cytotoxic, acidic phospholipase A2 purified from Indian cobra (Naja naja) venom: inhibition of anticoagulant activity by low molecular weight heparin.
Anticoagulant mechanism and platelet deaggregation property of a non-cytotoxic, acidic phospholipase A2 purified from Indian cobra (Naja naja) venom: inhibition of anticoagulant activity by low molecular weight heparin.
Biochimie (2015-01-13)
Sumita Dutta, Debananda Gogoi, Ashis K Mukherjee
ABSTRACT
In the present study, anticoagulant and platelet modulating activities of an acidic phospholipase A2 (NnPLA2-I) purified from Indian cobra Naja naja venom was investigated. The NnPLA2-I displayed a mass of 15.2 kDa and 14,186.0 Da when analyzed by SDS-PAGE and MALDI-TOF-MS, respectively. Peptide mass fingerprinting analysis of the NnPLA2-I showed its significant similarity with phospholipase A2 enzymes purified from cobra venom. BLAST analysis of one tryptic peptide sequence of NnPLA2-I demonstrated putative conserved domains of the PLA2-like superfamily. The Km and Vmax values of NnPLA2-I toward hydrolysis of its most preferred substrate-phosphotidylcholine (PC)-were determined to be 0.72 mM and 29.3 μmol min(-1) mg(-1), respectively. The anticoagulant activity of NnPLA2-I was found to be higher than the anticoagulant activity of heparin/AT-III or warfarin. The histidine modifying reagent, monovalent and polyvalent antivenom differentially inhibited the catalytic and anticoagulant activities of NnPLA2-I. Low molecular weight heparin did not inhibit the catalytic and platelet deaggregation activity of NnPLA2-I, albeit its anticoagulant activity was significantly reduced. The NnPLA2-I showed a non-enzymatic, mixed inhibition of thrombin with a Ki value of 9.3 nM. Heparin significantly decreased, with an IC50 value of 15.23 mIU, the thrombin inhibitory activity of NnPLA2-I. The NnPLA2-I uniquely increased the amidolytic activity of FXa without influencing its prothrombin activating property. NnPLA2-I showed dose-dependent deaggregation of platelet rich plasma (PRP) and inhibited the collagen and thrombin-induced aggregation of PRP. However, deaggregation of washed platelets by NnPLA2-I demonstrated in presence of PC or platelet poor plasma. Alkylation of histidine residue of NnPLA2-I resulted in 95% and 21% reduction of its platelet deaggregation and platelet binding properties, respectively. NnPLA2-I did not show cytotoxicity against human glioblastoma U87MG cells, bactericidal or hemolytic activity. The future therapeutic application of NnPLA2-I for treatment and prevention of cardiovascular disorders is therefore suggested.
Product Description
Supelco
Trifluoroacetic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitroaniline, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Pricing and availability is not currently available.
Supelco
4-Nitroaniline, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Pricing and availability is not currently available.