Merck
  • Functional evaluation using multifocal electroretinogram after selective retina therapy with a microsecond-pulsed laser.
Functional evaluation using multifocal electroretinogram after selective retina therapy with a microsecond-pulsed laser.
Investigative ophthalmology & visual science (2014-12-17)
Hoon Dong Kim, Jung Woo Han, Young-Hoon Ohn, Ralf Brinkmann, Tae Kwann Park
ABSTRACT
To evaluate the changes of retinal function with multifocal electroretinogram (mfERG), and estimate the association between functional and structural changes after selective retina therapy (SRT) with microsecond-pulsed laser in comparison to continuous wave laser photocoagulation (cwPC). Selective retina therapy and cwPC were applied with 10 × 10 shots and 1/2 lesion-width on the retina in the right and left eyes of 20 healthy Chinchilla Bastard rabbits, respectively. Optical coherence tomography (OCT), fundus fluorescein angiography (FFA), and mfERG were performed before, and on days 1, 7, and 30 after both laser treatments. The mean ratios of amplitudes and implicit times of N1 and P1 from eight hexagons covering laser-treated retinal lesions/total retina were measured. Histology was obtained after killing three rabbits at each time period to observe the anatomic changes after both laser treatments. The mean ratios of amplitudes of N1 and P1 in SRT lesions did not change significantly for 30 days after laser treatment. Only subtle reductions of the mean ratios of N1 and P1 amplitudes on day 1, thereafter the amplitudes showed the trend to recover toward baseline values. Histology and OCT revealed temporary and reversible morphologic changes after SRT, which restored to normal within 1 month. However, the mean ratios of N1 amplitudes on days 7 and 30 (P = 0.010, P < 0.001, respectively), and P1 amplitudes on days 7 and 30 (P < 0.001, P < 0.001, respectively) declined significantly in cwPC lesions compared with baseline. Disorganization and atrophic changes were identified on histology and OCT after cwPC. The results suggest that SRT preserved retinal function as well as anatomical structure after treatment.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Phenylephrine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Xylazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane