Merck
  • Synthesis and characterization of copolyanhydrides of carbohydrate-based galactaric acid and adipic acid.
Synthesis and characterization of copolyanhydrides of carbohydrate-based galactaric acid and adipic acid.
Carbohydrate research (2014-12-17)
Tuomas Mehtiö, Leena Nurmi, Virpi Rämö, Hannu Mikkonen, Ali Harlin
ABSTRACT
A series of copolyanhydrides, consisting of 2,3,4,5-tetra-O-acetylgalactaric acid (AGA) and adipic acid (AA) as monomer units, was polymerized. Synthesis of AGA monomer consisted of two steps. First, O-acetylation of galactaric acid secondary hydroxyl groups was performed using acetic anhydride as a reagent. Acetic anhydride was then further used as a reagent in the synthesis of diacetyl mixed anhydride of AGA. Polymerizations were conducted as bulk condensation polymerization at 150 °C. Thermal properties of the copolymers varied depending on monomer composition. Increase in the AGA content had a clear increasing effect on the Tg. A similar increasing effect was observed in Tm. The degree of crystallinity decreased as AGA content increased. There was a slightly lowering tendency in the molecular weights of the obtained polymers when the AGA content in the polymerization mixtures increased. The described synthesis route shows that bio-based aldaric acid monomers are potential candidates for the adjustment of thermal properties of polyanhydrides.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Mucic acid, 97%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard