Merck
  • Structural characterization of steroidal saponins from Smilax trinervula using ultra high-performance liquid chromatography coupled with LTQ-Orbitrap mass spectrometry.
Structural characterization of steroidal saponins from Smilax trinervula using ultra high-performance liquid chromatography coupled with LTQ-Orbitrap mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Yonghong Liang, Shun Yao, Fang Liang, Jian Liang, Guangyu Huang, Min Liu, Huilian Huang
ABSTRACT
Steroid saponins are a class of naturally existing substances widely distributed in the plants of Smilacaceae. Their biological activities have been attracting the interest of scientists in the chemical field for the past few years. To our best knowledge, there has been no study on structural characterization of steroidal saponins from Smilax trinervula (S. trinervula) using LTQ-Orbitrap mass spectrometry, which could provide an excellent approach for rapid screening of steroidal saponins in other plants of Smilacaceae. An ultra high-performance liquid chromatography coupled with high-resolution mass spectrometry (UHPLC/HRMS) method was therefore developed to characterize steroidal saponins in S. trinervula. This method was operated in both negative and positive ion modes with HRMS, as a result, a total of twenty-two steroidal saponins with three aglycone skeletons were elucidated in the crude extract from the root of S. trinervula. The characteristic-fragment ions could well identify and differentiate the three types of aglycone skeletons (diosgenyl saponins, furostanol saponins, C27-hydroxy diosgenyl saponins).
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Protodioscin, from Dioscorea spp., ≥98% (HPLC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%