Merck
  • Simultaneous determination of eperisone hydrochloride and paracetamol in mouse plasma by high performance liquid chromatography-photodiode array detector.
Simultaneous determination of eperisone hydrochloride and paracetamol in mouse plasma by high performance liquid chromatography-photodiode array detector.
Journal of chromatography. A (2015-02-24)
Marcello Locatelli, Roberta Cifelli, Cristina Di Legge, Renato Carmine Barbacane, Nicola Costa, Massimo Fresta, Christian Celia, Carlo Capolupo, Luisa Di Marzio
ABSTRACT
This paper reports the validation of a quantitative high performance liquid chromatography-photodiode array (HPLC-PDA) method for the simultaneous analysis, in mouse plasma, of eperisone hydrochloride and paracetamol by protein precipitation using zinc sulphate-methanol-acetonitrile. The analytes were resolved on a Gemini C18 column (4.6 mm × 250 mm; 5 μm particle size) using a gradient elution mode with a run time of 15 min, comprising re-equilibration, at 60°C (± 1°C). The method was validated over the concentration range from 0.5 to 25 μg/mL for eperisone hydrochloride and paracetamol, in mouse plasma. Ciprofloxacin was used as Internal Standard. Results from assay validations show that the method is selective, sensitive and robust. The limit of quantification of the method was 0.5 μg/mL for eperisone hydrochloride and paracetamol, and matrix-matched standard curves showed a good linearity, up to 25 μg/mL with correlation coefficients (r(2))≥ 0.9891. In the entire analytical range the intra and inter-day precision (RSD%) values were ≤ 1.15% and ≤ 1.46% for eperisone hydrochloride, and ≤ 0.35% and ≤ 1.65% for paracetamol. For both analytes the intra and inter-day trueness (bias%) values ranged, respectively, from -5.33% to 4.00% and from -11.4% to -4.00%. The method was successfully tested in pharmacokinetic studies after oral administration in mouse. Furthermore, the application of this method results in a significant reduction in terms of animal number, dosage, and improvement in speed, rate of analysis, and quality of pharmacokinetic parameters related to serial blood sampling.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis