Merck
  • Two jacalin-related lectins from seeds of the African breadfruit (Treculia africana L.).
Two jacalin-related lectins from seeds of the African breadfruit (Treculia africana L.).
Bioscience, biotechnology, and biochemistry (2014-08-27)
Michiko Shimokawa, Shadrack Makuta Nsimba-Lubaki, Namiko Hayashi, Yuji Minami, Fumio Yagi, Keiko Hiemori, Hiroaki Tateno, Jun Hirabayashi
ABSTRACT
Two jacalin-related lectins (JRLs) were purified by mannose-agarose and melibiose-agarose from seeds of Treculia africana. One is galactose-recognizing JRL (gJRL), named T. africana agglutinin-G (TAA-G), and another one is mannose-recognizing JRL (mJRL), TAA-M. The yields of the two lectins from the seed flour were approximately 7.0 mg/g for gJRL and 7.2 mg/g for mJRL. The primary structure of TAA-G was determined by protein sequencing of lysyl endopeptic peptides and chymotryptic peptides. The sequence identity of TAA-G to other gJRLs was around 70%. Two-residue insertion was found around the sugar-binding sites, compared with the sequences of other gJRLs. Crystallographic studies on other gJRLs have shown that the primary sugar-binding site of gJRLs can accommodate Gal, GalNAc, and GalNAc residue of T-antigen (Galβ1-3GalNAcα-). However, hemagglutination inhibition and glycan array showed that TAA-G did not recognize GalNAc itself and T-antigen. TAA-G preferred melibiose and core 3 O-glycan.
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, ≥99%, suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Melibiose, ≥98% (HPLC)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)