Merck
  • The adjuvant effect induced by di-(2-ethylhexyl) phthalate (DEHP) is mediated through oxidative stress in a mouse model of asthma.
The adjuvant effect induced by di-(2-ethylhexyl) phthalate (DEHP) is mediated through oxidative stress in a mouse model of asthma.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-06-24)
Huihui You, Shaohui Chen, Lin Mao, Bing Li, Ye Yuan, Rui Li, Xu Yang
ABSTRACT
Di-(2-ethylhexyl) phthalate, as the most commonly used plasticizer, is considered to be related to the asthma prevalence. There are studies affirming that the DEHP has an adjuvant effect in the pathogenesis of allergy asthma. Oxidative stress is one possible pathway for DEHP-adjuvant effect. Thus, this study explored whether DEHP could induce adjuvant effect in mouse asthma model via oxidative stress pathway. Male BALB/c mice were randomly divided into six groups: (1) saline group, (2) DEHP group, (3) ovalbumin (OVA) group, (4) DEHP+OVA group, (5) OVA+vitamin E (Vit E) group, (6) DEHP+OVA+Vit E group. The exposure dose of DEHP was 30 mg/kg body weight (bw)/day. After 18 days of the exposure protocol. Reactive oxygen species (ROS), glutathione (GSH) and malonaldehyde (MDA) levels and biomarkers related to asthma model were measured. Collectively, these data indicated higher ROS and MDA levels and lower GSH contents in DEHP+OVA group than that in OVA group, while Vit E, an antioxidant, could restore ROS, MDA and GSH levels to control levels and attenuate the DEHP and/or OVA effects. Our observations suggested that there was a relationship between oxidative stress and the adjuvant effect induced by DEHP in this mouse asthma model.
Sigma-Aldrich
Acetyl-β-methylcholine chloride, ≥98% (TLC), powder
Sigma-Aldrich
Rat IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Cotton Rat IL4 / Interleukin 4 ELISA Kit