Merck
WT1 interacts with MAD2 and regulates mitotic checkpoint function.
Nature communications (2014-09-19)
Jayasha Shandilya, Eneda Toska, Derek J Richard, Kathryn F Medler, Stefan G E Roberts
ABSTRACT
Tumour suppressors safeguard the fidelity of the mitotic checkpoint by transcriptional regulation of genes that encode components of the mitotic checkpoint complex (MCC). Here we report a new role for the tumour suppressor and transcription factor, WT1, in the mitotic checkpoint. We show that WT1 regulates the MCC by directly interacting with the spindle assembly checkpoint protein, MAD2. WT1 colocalizes with MAD2 during mitosis and preferentially binds to the functionally active, closed-conformer, C-MAD2. Furthermore, WT1 associates with the MCC containing MAD2, BUBR1 and CDC20, resulting in prolonged inhibition of the anaphase-promoting complex/cyclosome (APC/C) and delayed degradation of its substrates SECURIN and CYCLIN B1. Strikingly, RNA interference-mediated depletion of WT1 leads to enhanced turnover of SECURIN, decreased lag time to anaphase and defects in chromosome segregation. Our findings identify WT1 as a regulator of the mitotic checkpoint and chromosomal stability.
Sigma-Aldrich
MISSION® esiRNA, targeting human WT1
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mad2l1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mxi1
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O