Merck
  • Antibiotics and renal branching morphogenesis: comparison of toxicities.
Antibiotics and renal branching morphogenesis: comparison of toxicities.
Pediatric research (2014-08-29)
Ruud R G Bueters, Lisanne J A Kusters, Annelies Klaasen, Lambertus P van den Heuvel, Michiel F Schreuder
ABSTRACT
Many premature born neonates receive antibiotic drugs to treat infections, which are applied during active nephrogenesis. We studied the impact of clinical concentrations of gentamicin and alternatives, ceftazidime and meropenem, on ureteric branching. Mice metanephroi were dissected at embryonic day 13 and cultured in media with or without various concentrations of gentamicin, ceftazidime, or meropenem. Zero and 24 h kidney size were assessed by surface area measurements, and the ureteric tree was visualized by whole mount staining and confocal microscopy. Branching was evaluated by counting and gene expression levels of Wt1, Sox9, Bmp7, Fgf8, and Gdnf were investigated. A concentration of 2,000 μmol/l ceftazidime impaired ureteric development. In addition, a 4.5-fold and a 2.5-fold downregulation was noted in Fgf8 and Gdnf, respectively. No adverse effects were noted after gentamicin or meropenem treatment. No relationship was noted between surface area expansion and ureteric bud formation, but surface area at explantation related to bud count after 24 h of culture. Ceftazidime, but not gentamicin or meropenem reduced ureteric branching in mice and suggest a role for Fgf8 and Gdnf in its mechanism. Metanephros surface area measurements can be used to reduce intra- and inter-litter variation.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ceftazidime hydrate, 90.0-105.0%, contains ~10% sodium carbonate as stabilizer
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
USP
Ceftazidime pentahydrate, United States Pharmacopeia (USP) Reference Standard
USP
Meropenem, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ceftazidime hydrate
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%